Order Entry
Switzerland
ContactUsLinkComponent
 
Organic reference standard, Lutein solution, analytical standard, Supelco®
  79766-2.5ML
 :  Merck
 :  
undefined
Organic reference standard, Lutein solution, analytical standard, Supelco®
  79766-2.5ML
 :  Merck
 :  

 

  • Pk:
    2,5 mL
  • Standards:
    Analytical standard
  • CAS No.:
    127-40-2
  • Description:
    Lutein solution, ≥95,0% (HPLC)
  • Formula:
    C₄₀H₅₆O₂
  • Storage temperature:
    –20 °C
  • Description:
    Lutein solution

 

 

Lutein is a carotenoid, found in human serum and is also present in retina and macula lenses.