Organic reference standard, Indigo carmine, analytical standard, Supelco®
Catalog # 73436-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:860-22-0
- Description:Indigo carmine, ≥97,0% (HPLC)
- Formula:C₁₆H₈N₂Na₂O₈S₂
- Description:Indigo carmine
- Form:Neat
Specifications
About this item
Indigo carmine is a synthetic indigoid dye, which is generally used as a food colouring agent. It is basically used to maintain the natural color of food during processing or storage and also to improve the color, texture and appearance of food products.
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.