Order Entry
Switzerland
ContactUsLinkComponent
Supelco® Organic Reference Standard, Glycitein
Supelco® Organic Reference Standard, Glycitein
Catalog # 43534-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Glycitein
Catalog # 43534-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    40957-83-3
  • Description:
    Glycitein
  • Storage conditions:
    2...8 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00016679
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    4′,7-Dihydroxy-6-methoxyisoflavone; Glycetein
  • Formula:
    C₁₆H₁₂O₅
  • Molecular weight:
    284,26
  • Flash point:
    Not applicable

Specifications

About this item