Supelco® Organic Reference Standard, Forskolin
Katalognr. 93049-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:66575-29-9
- Description:Forskolin
- Storage conditions:2...30 °C
- Hazard class description:H312
- Shipping conditions:STD
- MDL No.:MFCD00082317
- M-Clarity quality level:MQ100
- Synonym(s):7β-Acetoxy-8,13-epoxy-1α,6β,9α-trihydroxylabd-14-en-11-one; Coleonol; Colforsin
- Molecular weight:410,50
Specifications
About this item
Caution: P280 - P302 + P352 + P312 - P362 + P364 - P501