Organic reference standard, Dimethyl sulfide, analytical standard, Supelco®
Catalog # 76048-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:75-18-3
- Description:Dimethyl sulfide, ≥98,0% (GC)
- Melting point:–98 °C (lit.)
- Formula:C₂H₆S
- Description:Dimethyl sulfide
- Flash point:–36 °C
- Boiling point:38 °C (lit.)
- Form:Neat
Specifications
About this item
Dimethyl sulfide is found to be the most abundant volatile thioether, which can find applications as a flavor enhancing agent in food. It can undergo oxidation in the process of malting and kilning, to yield dimethyl sulfoxide (DMSO).
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.