Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standard, Deoxynivalenol-¹³C₁₅ solution, analytical standard, Supelco®
Organic reference standard, Deoxynivalenol-¹³C₁₅ solution, analytical standard, Supelco®
Catalog # 34128-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Deoxynivalenol-¹³C₁₅ solution, analytical standard, Supelco®
Catalog # 34128-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    25 µg/ml in acetonitrile
  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    911392-36-4
  • Description:
    Deoxynivalenol-13C15 solution
  • M-Clarity quality level:
    MQ100
  • Formula:
    ¹³C₁₅H₂₀O₆
  • Storage temperature:
    –20 °C
  • Description:
    Deoxynivalenol-13C15 solution

Specifications

About this item

Deoxynivalenol-¹³C₁₅ (¹³C₁₅-DON) is a fusarium mycotoxin which belongs to trichothecene class of compounds.

¹³C₁₅-DON may be used as an internal standard for the identification of DON in maize by high performance liquid chromatography (HPLC) electrospray ionisation (ESI) mass spectrometry (MS/MS). Cereals and cereal-based compound animal feeds by LC-MS/MS operating on selected reaction monitoring (SRM) mode. Bioaerosols collected from Serpula lacrymans damaged homes by HPLC-MS/MS in multiple reaction monitoring (MRM) mode. It may also be used as an internal standard for the determination of HT-2 toxin, nivalenol and patulin in bioaerosols obtained from dairy cattle shed by HPLC-MS/MS.