Organic reference standard, Deoxynivalenol solution, analytical standard, Supelco®
Catalog # 34124-2ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in acetonitrile
- Pk:2 mL
- Standards:Analytical standard
- CAS No.:51481-10-8
- Description:Deoxynivalenol solution
- M-Clarity quality level:MQ100
- Formula:C₁₅H₂₀O₆
- Storage temperature:–20 °C
- Description:Deoxynivalenol solution
Specifications
About this item
Deoxynivalenol belongs to the group of tetracyclic sesquiterpenoid mycotoxins called trichothecenes, produced by Fusarium fungi, naturally occurring in small grains and infected corn.