Organic reference standard, Deoxynivalenol 3-glucoside solution, analytical standard, Supelco®
N° de catalogue 32911-1ML
Fournisseur: Merck
Numéro CAS:
Spécifications
- Concentration:50 µg/ml in acetonitrile
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:131180-21-7
- Description:Deoxynivalenol 3-glucoside solution
- Formula:C₂₁H₃₀O₁₁
- Storage temperature:–20 °C
- Description:Deoxynivalenol 3-glucoside solution
Spécifications
À propos de cet article
Deoxynivalenol 3-glucoside solution is the major metabolite of deoxynivalenol (DON) and a modified mycotoxin. The conjugation of deoxynivalenol (DON) with glucose formed by the metabolism of plants results in the formation of deoxynivalenol-3-glucoside.
Deoxynivalenol 3-glucoside may be used as a reference standard for the determination of deoxynivalenol 3-glucoside in wheat and barley by liquid chromatography coupled to mass spectrometry.