Supelco® Organic Reference Standard, Demethoxycurcumin
Catalog # 90593-10MG
Anbieter: Merck
CAS-Nummer:
Technische Daten
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:22608-11-3
- Description:Demethoxycurcumin
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD03427310
- M-Clarity quality level:MQ100
- Synonym(s):(E,E)-1-(4-Hydroxy-3-methoxyphenyl)-7-(4-hydroxyphenyl)-1,6-heptadiene-3,5-dione; Curcumin II; Desmethoxycurcumin; Monodemethoxycurcumin
- Molecular weight:338,35
Technische Daten
Über diesen Artikel
- GHS signal: GHS09