Supelco® Organic Reference Standard, D-(−)-Fructose
Catalog # F2793-500MG
Supplier: Merck
CAS Number:
Specifications
- Pk:500 mg
- CAS No.:57-48-7
- Description:D-(−)-Fructose
- Environmentally Preferable:
- Storage conditions:RT
- Shipping conditions:STD
- MDL No.:MFCD00148910
- M-Clarity quality level:MQ100
- Synonym(s):ᴅ-Levulose; Fruit sugar
- Melting point:119 - 122 °C (dec.) (lit.)
- Molecular weight:180,16