Order Entry
Switzerland
ContactUsLinkComponent
Supelco® Organic Reference Standard, Curcumin
Supelco® Organic Reference Standard, Curcumin
Codice catalogo 08511-10MG
Nome del fornitore:  Merck
Numero CAS:  
undefined
Supelco® Organic Reference Standard, Curcumin
Codice catalogo 08511-10MG
Nome del fornitore:  Merck
Numero CAS:  

Specifiche

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    458-37-7
  • Description:
    Curcumin
  • Storage conditions:
    2...8 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00008365
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (E,E)-1,7-bis(4-Hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione; Diferuloylmethane; Diferulylmethane; Natural Yellow 3
  • Formula:
    [HOC₆H₃(OCH₃)CH=CHCO]₂CH₂
  • Molecular weight:
    368,38
  • Storage temperature:
    2...8 °C
  • Flash point:
    Not applicable
  • Form:
    Powder

Specifiche

Informazioni su questo articolo

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.