Organic reference standard, Citral, analytical standard, Supelco®
Catalog # 15861-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:5392-40-5
- Description:Citral, ≥98,0% (sum of cis and trans, GC), 48,0 - 52,0% (cis), 48,0 - 52,0% (trans)
- Formula:C₁₀H₁₆O
- Storage temperature:2...8 °C
- Description:Citral
- Boiling point:229 °C(lit.)