Order Entry
Switzerland
ContactUsLinkComponent
Supelco® Organic Reference Standard, Chlorophyll B
Supelco® Organic Reference Standard, Chlorophyll B
  00538-1MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, Chlorophyll B
  00538-1MG
 :  Merck
 :  

 

  • Pk:
    1 mg
  • Standards:
    Analytical standard
  • CAS No.:
    519-62-0
  • Description:
    Chlorophyll b
  • Storage conditions:
    –20 °C
  • Shipping conditions:
    DICE
  • MDL No.:
    MFCD00079053
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₅₅H₇₀MgN₄O₆
  • Molecular weight:
    907,47
  • Storage temperature:
    −20 °C
  • Flash point:
    Not applicable