Supelco® Organic Reference Standard, Chlorophyll a
Catalog # 96145-1MG
Anbieter: Merck
CAS-Nummer:
Technische Daten
- Pk:1 mg
- Standards:Analytical standard
- CAS No.:479-61-8
- Description:Chlorophyll a
- Storage conditions:–20 °C
- Shipping conditions:DICE
- MDL No.:MFCD00079050
- M-Clarity quality level:MQ100
- Molecular weight:893,49