Supelco® Organic Reference Standard, Capsanthin
Catalog # 94863-1MG
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mg
- Standards:Analytical standard
- CAS No.:465-42-9
- Description:Capsanthin
- Storage conditions:–20 °C
- Shipping conditions:DICE
- M-Clarity quality level:MQ100
- Synonym(s):(3R,3′S,5′R)-3,3′-Dihydroxy-β,κ-caroten-6′-one
- Molecular weight:584,87