Organic reference standard, Brilliant Black BN, analytical standard, Supelco®
Katalognr. 11220-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:2519-30-4
- Description:Brilliant Black BN, ≥96,0% (HPLC)
- Formula:C₂₈H₁₇N₅Na₄O₁₄S₄
- Description:Brilliant Black BN
- Form:Neat
Specifications
About this item
Brilliant Black BN (BB) is a azo dye which is basically a tetra sodium salt. It is a synthetic dye mainly used as food additive. It is used as colouring agent to enhance the appearance. It is usually used in jellies, brown sauces and cake mixes.
- Used as colourant during quantification of synthetic colourants in fish roe and caviar for effective isolation using HPLC methods
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.