Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standard, Bisphenol A (2,3-dihydroxypropyl) glycidyl ether, analytical standard, Supelco®
Organic reference standard, Bisphenol A (2,3-dihydroxypropyl) glycidyl ether, analytical standard, Supelco®
Supplier:  Merck
Certificates
undefined
Organic reference standard, Bisphenol A (2,3-dihydroxypropyl) glycidyl ether, analytical standard, Supelco®
Supplier:  Merck

Specifications

  • CAS No.:
    76002-91-0
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₂₁H₂₆O₅
  • Storage temperature:
    2...8 °C
  • Description:
    Bisphenol A (2,3-dihydroxypropyl) glycidyl ether

Specifications

Frequently Bought Together