Supelco® Organic Reference Standard, Biochanin A
Catalog # 92142-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:491-80-5
- Description:Biochanin A
- Environmentally Preferable:
- Storage conditions:RT
- Shipping conditions:STD
- MDL No.:MFCD00006839
- M-Clarity quality level:MQ100
- Synonym(s):4′-Methylgenistein; 5,7-Dihydroxy-4′-methoxyisoflavone; Genistein 4′-methyl ether; Olmelin
- Melting point:210 - 213 °C (lit.)
- Molecular weight:284,26