Supelco® Organic Reference Standard, β-Tocotrienol
05644-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:490-23-3
- Description:β-Tocotrienol
- Storage conditions:–20 °C
- Shipping conditions:DICE
- MDL No.:MFCD09264634
- M-Clarity quality level:MQ100
- Synonym(s):(R)-β-Tocotrienol; [R-(E,E)]-3,4-Dihydro-2,5,8-trimethyl-2-(4,8,12-trimethyl-3,7,11-tridecatrienyl)-2H-1-benzopyran-6-ol
- Formula:C₂₈H₄₂O₂
- Molecular weight:410,63
- Storage temperature:−20 °C
- Flash point:Not applicable