Supelco® Organic Reference Standard, Bergamottin
Catalog # 01338-5MG-F
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mg
- Standards:Analytical standard
- CAS No.:7380-40-7
- Description:Bergamottin
- Storage conditions:2...8 °C
- Shipping conditions:STD
- MDL No.:MFCD00017409
- M-Clarity quality level:MQ100
- Synonym(s):Bergamotine; Bergaptin
- Melting point:75...80 °C
- Formula:C₂₁H₂₂O₄
- Molecular weight:338,40
- Storage temperature:2...8 °C
- Flash point:Not applicable
Specifications
About this item
Packaging: Bottomless glass bottle contents are inside inserted with fused cone.