Order Entry
Switzerland
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Aucubin
 :  Merck
 
undefined
Supelco® Organic Reference Standard, Aucubin
 :  Merck

 

  • CAS No.:
    479-98-1
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H302
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00136026
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Aucuboside; Rhimantin
  • Formula:
    C₁₅H₂₂O₉
  • Molecular weight:
    346,33
  • Storage temperature:
    2...8 °C
  • Flash point:
    Not applicable

 

Frequently Bought Together