Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standard, Aflatoxin M1 solution, analytical standard, Supelco®
Organic reference standard, Aflatoxin M1 solution, analytical standard, Supelco®
Catalog # 34031-2ML-R
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Aflatoxin M1 solution, analytical standard, Supelco®
Catalog # 34031-2ML-R
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    0.5 µg/ml in acetonitrile
  • Pk:
    2 mL
  • Standards:
    Analytical standard
  • CAS No.:
    6795-23-9
  • Description:
    Aflatoxin M1 solution
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₁₇H₁₂O₇
  • Storage temperature:
    –20 °C
  • Description:
    Aflatoxin M1 solution

Specifications

About this item

Isoproturon-d₆ has been used as an internal standard in the analysis of pesticides in fine airborne particulate matter, using liquid extraction under pressure and liquid chromatographic–tandem mass spectrometric method (HPLC-MS-MS).