SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Standards
Organic Standards
Organic reference standard, Aflatoxin G2, Supelco®
Organic reference standard, Aflatoxin G2, Supelco®
32757-5MG
:
Merck
:
Organic reference standard, Aflatoxin G2, Supelco®
32757-5MG
:
Merck
:
Pk:
5 mg
CAS No.:
7241-98-7
Description:
Aflatoxin G2
Formula:
C₁₇H₁₄O₇
Storage temperature:
–20 °C
Description:
Aflatoxin G2
Form:
Neat
:
Possibly carcinogenic.