Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standard, 5-Ring NOGE (Novolac glycidyl ether) mixture of isomers, chain-like or branched, analytical standard, Supelco®
Organic reference standard, 5-Ring NOGE (Novolac glycidyl ether) mixture of isomers, chain-like or branched, analytical standard, Supelco®
Catalog # 12109-50MG-F
Supplier:  Merck
CAS Number:  
Organic reference standard, 5-Ring NOGE (Novolac glycidyl ether) mixture of isomers, chain-like or branched, analytical standard, Supelco®
Catalog # 12109-50MG-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Analytical standard
  • Description:
    5-Ring NOGE (Novolac glycidyl ether) mixture of isomers, ≥90,0% (HPLC)
  • M-Clarity quality level:
    MQ100
  • Storage temperature:
    2...8 °C
  • Description:
    5-Ring NOGE (Novolac glycidyl ether) mixture of isomers
  • Form:
    Neat

Specifications

About this item