Organic reference standard, 5-(Hydroxymethyl)furfural, analytical standard, Supelco®
53407-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:67-47-0
- Description:5-(Hydroxymethyl)furfural, ≥98,0% (GC/HPLC)
- Melting point:28...34 °C (lit.)
- Formula:C₆H₆O₃
- Storage temperature:2…8 °C
- Description:5-(Hydroxymethyl)furfural
- Flash point:79 °C
- Boiling point:114...116 °C/1 mm Hg (lit.)
- Form:Neat
5-(Hydroxymethyl)furfural has been used as standard in the high performance liquid chromatographic (HPLC) analysis for the determination of 5-(hydroxymethyl)furfural content in co-hydrolysates and fermentation broth of lignocellulosic biomass for the lipid accumulation in M. isabellina type of microorganisms.
: Bottomless glass bottle. Contents are inside inserted fused cone.