Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standard, 4-Hydroxy-4-methyl-2-pentanone, analytical standard, Supelco®
Catalog # 78891-5ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 4-Hydroxy-4-methyl-2-pentanone, analytical standard, Supelco®
Catalog # 78891-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • CAS No.:
    123-42-2
  • Description:
    4-Hydroxy-4-methyl-2-pentanone, ≥98,5% (GC)
  • Seq:
    02
  • Formula:
    C₆H₁₂O
  • Description:
    4-Hydroxy-4-methyl-2-pentanone
  • Boiling point:
    166 °C (lit.)
  • Form:
    Neat

Specifications

About this item

4-Hydroxy-4-methyl-2-pentanone may be used as an internal standard in the analysis of volatile odor-active compounds present in wine using solid phase extraction with gas chromatography (SPE-GC).