Organic reference standard, 4-Butylresorcinol, analytical standard, Supelco®
49797-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:18979-61-8
- Description:4-Butylresorcinol, ≥97,0% (GC)
- Formula:C₁₀H₁₄O₂
- Description:4-Butylresorcinol
4-n-butylresorcinol is a derivative of resorcinol and a potent human tyrosinase inhibitor. It may be used to decrease skin irritation and is also known to inhibit melanin production.