Order Entry
Switzerland
ContactUsLinkComponent
 
Organic reference standard, 4-(4-Hydroxy-4-methylpentyl)-3-cyclohexene-1-carboxaldehyde, mixture of isomers, analytical standard, Supelco®
Catalog # 55862-10MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 4-(4-Hydroxy-4-methylpentyl)-3-cyclohexene-1-carboxaldehyde, mixture of isomers, analytical standard, Supelco®
Catalog # 55862-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    31906-04-4
  • Description:
    4-(4-Hydroxy-4-methylpentyl)-3-cyclohexene-1-carboxaldehyde, mixture of isomers, ≥95,0% (sum of isomers, GC); 15,0 - 35,0% (minor component, GC); 65,0 - 85,0% (GC)
  • Formula:
    C₁₃H₂₂O₂
  • Storage temperature:
    2...8 °C
  • Description:
    4-(4-Hydroxy-4-methylpentyl)-3-cyclohexene-1-carboxaldehyde
  • Flash point:
    93 °C
  • Form:
    Neat

Specifications

About this item