Organic reference standard, 3-Methyl-5-(2,2,3-trimethyl-3-cyclopenten-1-yl)-4-penten-2-ol, analytical standard, Supelco®
Catalog # 44064-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:Analytical standard
- CAS No.:67801-20-1
- Description:3-Methyl-5-(2,2,3-trimethyl-3-cyclopenten-1-yl)-4-penten-2-ol, ≥90% (GC)
- Formula:C₁₄H₂₄O
- Description:3-Methyl-5-(2,2,3-trimethyl-3-cyclopenten-1-yl)-4-penten-2-ol