Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standard, 2,3-Hexanedione, analytical standard, Supelco®
Organic reference standard, 2,3-Hexanedione, analytical standard, Supelco®
Catalog # 75034-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 2,3-Hexanedione, analytical standard, Supelco®
Catalog # 75034-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    3848-24-6
  • Description:
    2,3-Hexanedione, ≥97,0% (GC)
  • Formula:
    C₆H₁₀O₂
  • Storage temperature:
    2...8 °C
  • Description:
    2,3-Hexanedione
  • Flash point:
    28 °C
  • Boiling point:
    128 °C (lit.)

Specifications

About this item

2,3-Hexanedione can be used as an internal standard for the quantitative determination of methylglyoxal present in the tissues of Sprague-Dawley rats using electrospray ionization liquid chromatography coupled with mass spectrometry(ESI/LC/MS).

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.