Supelco® Organic Reference Standard, [10]-Gingerol
Catalog # 42630-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:23513-15-7
- Description:[10]-Gingerol
- Storage conditions:2...8 °C
- Hazard class description:H315 - H319 - H335
- Shipping conditions:STD
- MDL No.:MFCD01682694
- M-Clarity quality level:MQ100
- Synonym(s):(S)- 5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)-3-tetradecanone; 10-Gingerol
- Formula:C₂₁H₃₄O₄
- Molecular weight:350,49
- Storage temperature:2…8 °C
- Flash point:166,3 °C - closed cup
- Form:Neat
Specifications
About this item
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.
Caution: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338