Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standard, (2S-cis)-(-)-5-Benzyl-3,6-dioxo-2-piperazineacetic acid, analytical standard, Supelco®
Organic reference standard, (2S-cis)-(-)-5-Benzyl-3,6-dioxo-2-piperazineacetic acid, analytical standard, Supelco®
Catalog # 89274-50MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, (2S-cis)-(-)-5-Benzyl-3,6-dioxo-2-piperazineacetic acid, analytical standard, Supelco®
Catalog # 89274-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Analytical standard
  • CAS No.:
    5262-10-2
  • Description:
    (2S-cis)-(-)-5-Benzyl-3,6-dioxo-2-piperazineacetic acid, ≥97,0% (HPLC)
  • Melting point:
    270...272 °C (lit.)
  • Formula:
    C₁₃H₁₄N₂O₄
  • Storage temperature:
    2…8 °C
  • Description:
    (2S-cis)-(-)-5-Benzyl-3,6-dioxo-2-piperazineacetic acid
  • Form:
    Neat

Specifications

About this item

Degradation product from aspartame (E951).

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.