Order Entry
Switzerland
ContactUsLinkComponent
Supelco® Organic Reference Standard, (±)-Citronellal
Catalog # 72638-1ML
Anbieter:  Merck
CAS-Nummer:  
undefined
Supelco® Organic Reference Standard, (±)-Citronellal
Catalog # 72638-1ML
Anbieter:  Merck
CAS-Nummer:  

Technische Daten

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    106-23-0
  • Description:
    (±)-Citronellal
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H315 - H317 - H319
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00038090
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (±)-3,7-Dimethyl-6-octenal
  • Formula:
    (CH₃)₂C=CHCH₂CH₂CH(CH₃)CH₂CHO
  • Molecular weight:
    154,25
  • Density:
    0,857 g/ml at 25 °C (lit.)
  • Flash point:
    74 °C - Closed cup
  • Boiling point:
    207 °C (lit.)

Technische Daten

Über diesen Artikel

  • GHS signal: GHS07, GHS09
Caution: P261 - P264 - P272 - P280 - P302 + P352 - P305 + P351 + P338