Organic reference standard, (-)-Rose oxide, analytical standard, Supelco®
(-)-Rose oxide, ≥99,0% (mixture of isomers (cis+trans), GC), Analytical standard, Neat, 1 mL
Catalog # 83917-1ML
Supplier: Merck
Organic reference standard, (-)-Rose oxide, analytical standard, Supelco®
Catalog # 83917-1ML
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1 mL
- Standards:Analytical standard
- Description:(-)-Rose oxide, ≥99,0% (mixture of isomers (cis+trans), GC)
- Form:Neat
- Cat. No.:83917-1ML
- M-Clarity quality level:MQ100
- Formula:C₁₀H₁₈O
- Description:(-)-Rose oxide
- CAS No.:16409-43-1



