Organic reference standard, (-)-Myrtenol, analytical standard, Supelco®
Catalog # 70158-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:19894-97-4
- Description:(-)-Myrtenol, ≥98,5% (sum of enantiomers, GC)
- Environmentally Preferable:
- M-Clarity quality level:MQ100
- Formula:C₁₀H₁₆O
- Storage temperature:2…8 °C
- Description:(-)-Myrtenol
- Boiling point:220...222 °C (lit.)
- Form:Neat
Specifications
About this item
Use as chiral building block.