Order Entry
Switzerland
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, (−)-Epigallocatechin
  08108-1MG-F
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, (−)-Epigallocatechin
  08108-1MG-F
 :  Merck
 :  

 

  • Pk:
    1 mg
  • Standards:
    Analytical standard
  • CAS No.:
    970-74-1
  • Description:
    (−)-Epigallocatechin
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00075939
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (−)-cis-2-(3,4,5-Trihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol; (−)-cis-3,3′,4′,5,5′,7-Hexahydroxyflavane
  • Formula:
    C₁₅H₁₄O₇
  • Molecular weight:
    306,27
  • Flash point:
    Not applicable