Order Entry
Switzerland
ContactUsLinkComponent
Supelco® Organic Reference Standard, (−)-Cotinine
Supelco® Organic Reference Standard, (−)-Cotinine
Catalog # 74003-25MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, (−)-Cotinine
Catalog # 74003-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    486-56-6
  • Description:
    (−)-Cotinine
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H302 - H315 - H319 - H335
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD00077696
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (S)-1-Methyl-5-(3-pyridyl)-2-pyrrolidinone; S(−)-1-Methyl-5-(3-pyridyl)-2-pyrrolidone
  • Melting point:
    40 - 42 °C (lit.)
  • Molecular weight:
    176,22
  • Flash point:
    110 °C - Closed cup
  • Boiling point:
    250 °C/150 mmHg (lit.)

Specifications

About this item

  • GHS signal: GHS07
Caution: P261 - P264 - P270 - P301 + P312 - P302 + P352 - P305 + P351 + P338