ContactUsLinkComponent
Pint bottle brush
  MICRBRWH-66
 :  MICRONOVA
undefined
Pint bottle brush
  MICRBRWH-66
 :  MICRONOVA
  • Brush head Ø:
    63,5 mm
  • Handle material:
    Plastic
  • Total length:
    305 mm
  • Brush head length:
    114 mm
  • Pk:
    1
This wirewound brush features a smaller diameter handle and softer bristles for delicate cleaning of jars and bottles with smaller openings.
  • BPA free
Base resins used to produce block and bristles are made from FDA approved materials.