Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Bottle brush

Bottle brush

178 mm, 76 mm, Plastic, 406 mm
Catalog # MICRBRWH-67
Supplier:   MICRONOVA
Bottle brush
Catalog # MICRBRWH-67
Supplier:  MICRONOVA
Bottle brush
Bottle brush

178 mm, 76 mm, Plastic, 406 mm

Price
SFr. 124.00
Log in for availability
1 * 1 ST
Product Details & Documents

This wirewound brush features a smaller diameter handle and softer bristles for delicate cleaning of jars and bottles with smaller openings.

  • BPA-free

Base resins used to produce block and bristles are made from FDA approved materials.

Specifications
  • Brush head Ø:
    76 mm
  • Handle material:
    Plastic
  • Total length:
    406 mm
  • Brush head length:
    178 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.