Order Entry
Switzerland
ContactUsLinkComponent
Hydroxystilbamidine bis(methanesulphonate) 4% in water fluorescent dye, for histology
Irritant
Hydroxystilbamidine bis(methanesulphonate) 4% in water fluorescent dye, for histology
Catalog # 80023.
Supplier:  Biotium
CAS Number:  
undefined
Irritant
Hydroxystilbamidine bis(methanesulphonate) 4% in water fluorescent dye, for histology
Catalog # 80023.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    200 µl
  • Seq:
    1358

Specifications

About this item

 :
 :472.54 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:223769-64-0
Formula:C₁₈H₂₄N₄O₇S₂
MDL Number:MFCD01863083