Order Entry
Switzerland
ContactUsLinkComponent
Calcein AM 1 mg/mL in anhydrous DMSO fluorescent dye, for histology
Calcein AM 1 mg/mL in anhydrous DMSO fluorescent dye, for histology
Catalog # 80011-2
Supplier:  Biotium
CAS Number:  
undefined
Calcein AM 1 mg/mL in anhydrous DMSO fluorescent dye, for histology
Catalog # 80011-2
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :994.86 g/mol
Storage Temperature:Freezer
CAS Number:148504-34-1
Formula:C₄₆H₄₆N₂O₂₃
MDL Number:MFCD05861516