Order Entry
Switzerland
ContactUsLinkComponent
DiOC₅(3) iodide membrane stain, for histology
DiOC₅(3) iodide membrane stain, for histology
Catalog # 70007.
Supplier:  Biotium
CAS Number:  
undefined
DiOC₅(3) iodide membrane stain, for histology
Catalog # 70007.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :544.47 g/mol
Storage Temperature:Freezer
CAS Number:53213-81-3
Formula:C₂₇H₃₃IN₂O₂