Order Entry
Switzerland
ContactUsLinkComponent
 
Tocopheryl acetate (Vitamin E acetate) solution, certified reference material, Supelco®
Catalog # V-033-1ML
Supplier:  Merck
CAS Number:  
Tocopheryl acetate (Vitamin E acetate) solution, certified reference material, Supelco®
Catalog # V-033-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1,0 mg/ml in methanol
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    52225-20-4
  • Description:
    Tocopheryl acetate (Vitamin E acetate) solution
  • Description:
    Tocopheryl acetate (Vitamin E acetate) solution

Specifications

About this item