Supelco® Cerilliant® Organic Reference Standard, Thiamine HCl (Vitamin B1) solution
V-014-1ML
: Merck
:
- Concentration:1,0 mg/ml in methanol (as free base)
- Pk:1 mL
- Standards:Certified reference material, Cerilliant®
- CAS No.:67-03-8
- Description:Thiamine HCl (Vitamin B1) solution
- Storage conditions:–20 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:WICE
- MDL No.:MFCD00012780
- M-Clarity quality level:MQ300
- Synonym(s):Thiamine hydrochloride; Vitamin B₁; Aneurine hydrochloride; Vitamin B₁ hydrochloride
- Melting point:250 °C (dec.) (lit.)
- Molecular weight:337,27
- Flash point:9,7 °C - Closed cup
- Pk Info:Ampule of 1 ml
- GHS signal: GHS02, GHS06, GHS08
: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311