Order Entry
Switzerland
ContactUsLinkComponent
Japanese aflatoxin mixture, certified reference material, Supelco®
Japanese aflatoxin mixture, certified reference material, Supelco®
Catalog # CRM40139
Supplier:  Merck
CAS Number:  
Japanese aflatoxin mixture, certified reference material, Supelco®
Catalog # CRM40139
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    25 µg/ml in acetonitrile
  • Pk:
    5
  • Standards:
    Certified reference material
  • Description:
    Japanese Aflatoxin Mixture
  • Pk Info:
    Ampoule of 5 x 1 ml

Specifications

About this item