Order Entry
Switzerland
ContactUsLinkComponent
DASPEI (2-(4-(Dimethylamino)styryl)-N-ethylpyridinium iodide) mitochondria dye, for fluorescence microscopy
Irritant
DASPEI (2-(4-(Dimethylamino)styryl)-N-ethylpyridinium iodide) mitochondria dye, for fluorescence microscopy
Katalognr. 70018.
Supplier:  Biotium
CAS Number:  
Irritant
DASPEI (2-(4-(Dimethylamino)styryl)-N-ethylpyridinium iodide) mitochondria dye, for fluorescence microscopy
Katalognr. 70018.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :380.273 g/mol
EINECS:223-248-3
 :GHS07
Storage Temperature:Refrigerator
CAS Number:3785-01-1
Formula:C₁₇H₂₁IN₂
MDL Number:MFCD00011994