Order Entry
Switzerland
ContactUsLinkComponent
DiIC1(5) iodide mitochondria dye, for fluorescence microscopy
DiIC1(5) iodide mitochondria dye, for fluorescence microscopy
Catalog # 70015.
Supplier:  Biotium
CAS Number:  
DiIC1(5) iodide mitochondria dye, for fluorescence microscopy
Catalog # 70015.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :510.5 g/mol
Storage Temperature:Refrigerator
CAS Number:36536-22-8
Formula:C₂₇H₃₁IN₂