Order Entry
Switzerland
ContactUsLinkComponent
Acridine Orange 10-nonyl bromide (NAO) mitochondria dye, for fluorescence microscopy
Irritant
Acridine Orange 10-nonyl bromide (NAO) mitochondria dye, for fluorescence microscopy
Catalog # 70012.
Supplier:  Biotium
CAS Number:  
Irritant
Acridine Orange 10-nonyl bromide (NAO) mitochondria dye, for fluorescence microscopy
Catalog # 70012.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :472.50 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:75168-11-5
Formula:C₂₆H₃₈BrN₃
MDL Number:MFCD00467605