Order Entry
Switzerland
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, QC-Standard Solution for EN ISO 9377-2
Catalog # 51706-1ML-F
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, QC-Standard Solution for EN ISO 9377-2
Catalog # 51706-1ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • Description:
    QC-Standard solution for EN ISO 9377-2
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H225 - H304 - H315 - H336 - H351 - H410
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Flash point:
    –18 °C – Closed cup

Specifications

About this item

  • GHS signal: GHS02, GHS08, GHS07, GHS09
Caution: P202 - P210 - P273 - P301 + P310 - P303 + P361 + P353 - P331