Poly-L-lysine hydrobromide
About this item
Poly-L-lysine is a positively charged amino acid polymer with approximately one HBr per lysine residue. The hydrobromide allows the poly-L-lysine to be in a crystalline form soluble in water. A small amount of product may be found in the beta structure because the HBr interferes with hydrogen bonding between amino and either the carboxyl groups or N or O containing moieties.
Poly-L-lysine polymers can be used in promoting cell adhesion to solid substrates, conjugation to methotrexate for increased drug transport, microencapsulation of islets, cell microencapsulation technology, microarray glass slide coating, and chromosomal preparations. Lower molecular weight poly-L-lysine (30,000-70,000) is less viscuous in solution, but higher molecular weight versions provide more attachment sites per molecule.
This product is supplied as a 0.10% (w/v) solution in water. Coating slides with this solution should be followed with a 5 minute incubation and drying the slides at room temperature. Coated slides will be stable for one year if protected from dust.
2 Options Available Below
Product Details & Documents
Poly-L-lysine is a positively charged amino acid polymer with approximately one HBr per lysine residue. The hydrobromide allows the poly-L-lysine to be in a crystalline form soluble in water. A small amount of product may be found in the beta structure because the HBr interferes with hydrogen bonding between amino and either the carboxyl groups or N or O containing moieties.
Poly-L-lysine polymers can be used in promoting cell adhesion to solid substrates, conjugation to methotrexate for increased drug transport, microencapsulation of islets, cell microencapsulation technology, microarray glass slide coating, and chromosomal preparations. Lower molecular weight poly-L-lysine (30,000-70,000) is less viscuous in solution, but higher molecular weight versions provide more attachment sites per molecule.
This product is supplied as a 0.10% (w/v) solution in water. Coating slides with this solution should be followed with a 5 minute incubation and drying the slides at room temperature. Coated slides will be stable for one year if protected from dust.
Documents
Merck The Color of Tissue Diagnostics
Product Literature
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- P8920-100ML
- P8920-500ML
- Protein/peptide name:
- Poly-L-lysine hydrobromide
- Poly-L-lysine hydrobromide
- CAS No.:
- 25988-63-0
- 25988-63-0
- MDL No.:
- MFCD00237230
- MFCD00237230
- Concentration:
- 0.1 % w/v
- 0.1 % w/v
- Formulation:
- Solution in water
- Solution in water
- Molecular weight:
- 150000 - 300000
- 150000 - 300000
- Molecular formula:
- (C₆H₁₃N₂OBr)n
- (C₆H₁₃N₂OBr)n
- Storage conditions:
- Ambient
- Ambient
- M-Clarity quality level:
- MQ200
- MQ200
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.P8920-100MLProtein/peptide namePoly-L-lysine hydrobromideCAS No.25988-63-0MDL No.MFCD00237230Concentration0.1 % w/vFormulationSolution in waterMolecular weight150000 - 300000Molecular formula(C₆H₁₃N₂OBr)nStorage conditionsAmbientM-Clarity quality levelMQ200 - Specifications:
More
Specifications
Cat. No.P8920-500MLProtein/peptide namePoly-L-lysine hydrobromideCAS No.25988-63-0MDL No.MFCD00237230Concentration0.1 % w/vFormulationSolution in waterMolecular weight150000 - 300000Molecular formula(C₆H₁₃N₂OBr)nStorage conditionsAmbientM-Clarity quality levelMQ200



